siRNA Details
Gene Symbol: Sepw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009156.2 | NP_033182.1 | 4 | 234 | ||
GenBank mRNA | AF015284.1 | - | 4 | 189 | ||
AF241527.2 | - | 4 | 187 | |||
AK002870.1 | - | 4 | 210 | |||
AK028124.1 | - | 4 | 204 | |||
BC052719.1 | - | 4 | 196 | 6814242 | ||
BU604713.1 | - | 4 | 267 | |||
U67890.1 | - | 4 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
