siRNA Details
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009161.4 | NP_033187.1 | 4 | 546 | ||
GenBank mRNA | AB024920.1 | - | 4 | 546 | ||
AF019564.1 | - | 3 | 293 | |||
AK009015.1 | - | 3 | 309 | |||
AK075915.1 | - | 3 | 318 | |||
BC060287.1 | - | 3 | 305 |
