siRNA Details
Gene Symbol: Sh2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011364.3 | NP_035494.1 | 2, 3 | 326 | ||
GenBank mRNA | AF072903.1 | - | 2, 3 | 373 | ||
AF097632.1 | - | 2, 3 | 272 | |||
AF097633.1 | - | 2, 3 | 265 | |||
AK040076.1 | - | 2, 3 | 327 | |||
AK042476.1 | - | 155 | ||||
BC028467.2 | - | 1 | 917 | |||
BC147367.1 | - | 2, 3 | 236 | 9056424 | ||
BC147368.1 | - | 2, 3 | 236 | 9056435 | ||
BC147807.1 | - | 2, 3 | 99 | 9087524 | ||
BC147811.1 | - | 2, 3 | 99 | 9087528 | ||
KF811030.1 | - | 3, 4 | 265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
