siRNA Details
Gene Symbol: Shd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159523.1 | NP_001152995.1 | 4 | 702 | ||
NM_009168.2 | NP_033194.1 | 4 | 699 | |||
GenBank mRNA | AB018423.1 | - | 4 | 698 | ||
AK142491.1 | - | 4 | 715 | |||
AK164639.1 | - | 4 | 635 | |||
BC103798.1 | - | 4 | 595 | 30288676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
