siRNA Details
Gene Symbol: Shfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009169.2 | NP_033195.1 | 3 | 385 | ||
GenBank mRNA | AK135462.1 | - | 338 | |||
AK160432.1 | - | 3 | 378 | |||
AK161690.1 | - | 3 | 358 | |||
AK167914.1 | - | 3 | 335 | |||
BC023490.1 | - | 3 | 342 | 5251089 | ||
BC058117.1 | - | 3 | 351 | 6412144 | ||
U41626.1 | - | 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
