siRNA Details
Gene Symbol: Siah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009174.3 | NP_033200.2 | 2 | 1267 | ||
GenBank mRNA | AK139626.1 | - | 3 | 848 | ||
AK149316.1 | - | 2 | 1282 | |||
BC058400.1 | - | 2 | 1267 | 5701212 | ||
Z19581.1 | - | 2 | 983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
