siRNA Details
Gene Symbol: St6galnac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011371.2 | NP_035501.2 | 4, 5 | 1056 | ||
GenBank mRNA | BC141095.1 | - | 1039 | 9055707 | ||
BC145373.1 | - | 1039 | 9053931 | |||
BC145374.1 | - | 4, 5 | 1006 | 9053932 | ||
Y11274.1 | - | 4, 5 | 1056 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
