siRNA Details
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035228.2 | NP_001030305.1 | 4 | 514 | ||
NM_011375.3 | NP_035505.2 | 4 | 726 | |||
GenBank mRNA | AB013302.1 | - | 3 | 265 | ||
AB018048.1 | - | 4 | 454 | |||
AF119416.1 | - | 4 | 504 | |||
AK012961.1 | - | 4 | 495 | |||
AK158594.1 | - | 4 | 548 | |||
AK165726.1 | - | 4 | 516 | |||
BC138557.1 | - | 4 | 431 | 8861577 | ||
BC138559.1 | - | 4 | 429 | 8861579 | ||
Y15003.1 | - | 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
