siRNA Details
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159529.1 | NP_001153001.1 | 4 | 643 | ||
NM_009187.3 | NP_033213.1 | 3 | 574 | |||
GenBank mRNA | AK009614.1 | - | 4 | 640 | ||
AK133520.1 | - | 3 | 574 | |||
AK150034.1 | - | 3 | 571 | |||
AK152553.1 | - | 3 | 564 | |||
AK159441.1 | - | 3 | 572 | |||
AK166761.1 | - | 3 | 559 | |||
AK167062.1 | - | 3 | 569 | |||
AK169619.1 | - | 3 | 592 | |||
AK198092.1 | - | 1 | 127 | |||
AK200674.1 | - | 1 | 127 | |||
AK208783.1 | - | 1 | 127 | |||
AK217840.1 | - | 1 | 127 | |||
BC004715.1 | - | 3 | 544 | 3582142 | ||
BC085296.1 | - | 3 | 549 | 5337317 | ||
X80899.1 | - | 3 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
