siRNA Details
Gene Symbol: Cox7a2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159529.1 | NP_001153001.1 | 2 | 160 | ||
NM_009187.3 | NP_033213.1 | 2 | 160 | |||
GenBank mRNA | AK009614.1 | - | 2 | 157 | ||
AK133520.1 | - | 2 | 160 | |||
AK150034.1 | - | 2 | 157 | |||
AK159441.1 | - | 2 | 158 | |||
AK166761.1 | - | 2 | 145 | |||
AK167062.1 | - | 2 | 155 | |||
AK169619.1 | - | 2 | 172 | |||
AK184659.1 | - | 1 | 35 | |||
AK216072.1 | - | 1 | 35 | |||
AK216357.1 | - | 1 | 35 | |||
BC004715.1 | - | 2 | 124 | 3582142 | ||
BC085296.1 | - | 2 | 129 | 5337317 | ||
BY122624.1 | - | 2 | 160 | |||
X80899.1 | - | 2 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
