siRNA Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110350.1 | NP_001103820.1 | 15, 16 | 3147 | ||
NM_001110351.1 | NP_001103821.1 | 16, 17 | 3024 | |||
NM_011378.2 | NP_035508.2 | 16, 17 | 3255 | |||
GenBank mRNA | AK032209.1 | - | 15, 16 | 3014 | ||
AK136347.1 | - | 10, 11 | 1949 | |||
AK136972.1 | - | 1, 2 | 380 | |||
AK220292.1 | - | 15, 16 | 3124 | |||
BC052716.1 | - | 15, 16 | 3034 | 6837144 | ||
BC053385.1 | - | 15, 16 | 3017 | |||
BC078454.1 | - | 15, 16 | 3008 | |||
L36831.1 | - | 15, 16 | 2938 | |||
L38620.2 | - | 14, 15 | 2914 | |||
L38621.2 | - | 14, 15 | 2914 | |||
U22394.1 | - | 16, 17 | 3263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
