siRNA Details
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009188.4 | NP_033214.2 | 11 | 1526 | ||
GenBank mRNA | AK087970.1 | - | 4 | 623 | ||
AK173005.1 | - | 10 | 3476 | |||
BC129859.1 | - | 11 | 1498 | |||
L38622.2 | - | 11 | 1498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
