siRNA Details
Gene Symbol: Six1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009189.3 | NP_033215.2 | 1, 2 | 849 | ||
GenBank mRNA | AK031122.1 | - | 1, 2 | 604 | ||
AK131925.1 | - | 1, 2 | 850 | |||
AK144753.1 | - | 1, 2 | 860 | |||
BC023304.1 | - | 1, 2 | 834 | 4188451 | ||
X80339.1 | - | 1, 2 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
