siRNA Details
Gene Symbol: Six3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011381.4 | NP_035511.2 | 1 | 875 | ||
GenBank mRNA | BC094426.1 | - | 678 | 6855602 | ||
BC098096.1 | - | 1 | 495 | 6402835 | ||
BF467709.1 | - | 1 | 513 | |||
D83144.1 | - | 1 | 1136 | |||
D83145.1 | - | 1 | 461 | |||
X90871.1 | - | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
