siRNA Details
Gene Symbol: Vps4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009190.2 | NP_033216.2 | 10 | 1404 | ||
GenBank mRNA | AK034641.1 | - | 62 | |||
AK151023.1 | - | 10 | 1358 | |||
AK152131.1 | - | 10 | 1391 | |||
AK165594.1 | - | 1349 | ||||
AK168270.1 | - | 10 | 1295 | |||
BC003799.1 | - | 10 | 1311 | 3488471 | ||
U10119.1 | - | 10 | 1355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
