siRNA Details
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011385.2 | NP_035515.2 | 4 | 1462 | ||
GenBank mRNA | AF435852.1 | - | 4 | 1441 | ||
AK019148.2 | - | 4 | 519 | |||
BC030841.1 | - | 1 | 239 | |||
BC068305.1 | - | 5 | 1306 | 30093626 | ||
CB722801.1 | - | 1 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
