siRNA Details
Gene Symbol: Skil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039090.2 | NP_001034179.1 | 3 | 1857 | ||
NM_001271772.1 | NP_001258701.1 | 5 | 1538 | |||
NM_011386.3 | NP_035516.2 | 3 | 1857 | |||
GenBank mRNA | AB221599.1 | - | 2 | 1134 | ||
AK154471.1 | - | 3 | 1804 | |||
AK171402.1 | - | 3 | 1806 | |||
BC018600.1 | - | 5 | 1470 | |||
BC049934.1 | - | 2 | 1283 | 5387475 | ||
U10530.1 | - | 2 | 1628 | |||
U10532.1 | - | 2 | 1628 | |||
U14655.1 | - | 2 | 1190 |
