siRNA Details
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205311.1 | NP_001192240.1 | 16 | 2022 | ||
NM_019415.2 | NP_062288.2 | 16 | 2019 | |||
GenBank mRNA | AK052691.1 | - | 16 | 2023 | ||
AK052755.1 | - | 16 | 2021 | |||
AK085496.1 | - | 16 | 2028 | |||
BC038612.1 | - | 16 | 2044 | 4237274 | ||
U61085.1 | - | 16 | 2001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
