siRNA Details
Gene Symbol: Slc12a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253804.1 | NP_001240733.1 | 18 | 2339 | ||
NM_009195.3 | NP_033221.1 | 18 | 2333 | |||
GenBank mRNA | AF047339.1 | - | 18 | 2257 | ||
AF121118.1 | - | 18 | 2324 | |||
AK149969.1 | - | 18 | 2322 | |||
AK151717.1 | - | 18 | 2294 | |||
AK152762.1 | - | 18 | 2311 | |||
AK153245.1 | - | 18 | 2311 | |||
AK159274.1 | - | 18 | 2321 | |||
AK159331.1 | - | 18 | 2316 | |||
AK159483.1 | - | 18 | 2289 | |||
AK159537.1 | - | 18 | 2316 | |||
AK160034.1 | - | 18 | 2282 | |||
AK160072.1 | - | 18 | 2316 | |||
AK164479.1 | - | 18 | 2298 | |||
AK165874.1 | - | 18 | 2311 | |||
AK166304.1 | - | 18 | 2292 | |||
AK167371.1 | - | 18 | 2330 | |||
AK167444.1 | - | 18 | 2330 | |||
AK169044.1 | - | 18 | 2271 | |||
BC066872.1 | - | 18 | 2263 | 30436525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
