siRNA Details
Gene Symbol: Slc16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009196.4 | NP_033222.1 | 4 | 1049 | ||
GenBank mRNA | AF058055.1 | - | 4 | 1030 | ||
AK033516.1 | - | 4 | 1047 | |||
AK053421.1 | - | 1 | 410 | |||
AK079379.1 | - | 4 | 1045 | |||
AK136198.1 | - | 1 | 42 | |||
AK161701.1 | - | 4 | 1047 | |||
AK162682.1 | - | 4 | 1046 | |||
AK166606.1 | - | 4 | 1047 | |||
BC014777.1 | - | 4 | 1057 | 3499773 | ||
X82438.1 | - | 3 | 841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
