siRNA Details
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009199.2 | NP_033225.1 | 3 | 386 | ||
GenBank mRNA | AK018188.1 | - | 3 | 419 | ||
AK045632.1 | - | 3 | 418 | |||
AK083097.1 | - | 3 | 419 | |||
BC064797.1 | - | 3 | 408 | 30029501 | ||
BC065099.1 | - | 3 | 410 | 30364069 | ||
D43797.1 | - | 3 | 392 | |||
U73521.1 | - | 3 | 386 |

More Information
Other Species Targeted

Gene Ontology Categories:
