siRNA Details
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077514.3 | NP_001070982.1 | 4 | 963 | ||
NM_001077515.2 | NP_001070983.1 | 4 | 390 | |||
NM_011393.2 | NP_035523.1 | 4 | 390 | |||
GenBank mRNA | AB007810.1 | - | 4 | 1020 | ||
AB007811.1 | - | 4 | 379 | |||
AB007812.1 | - | 4 | 390 | |||
AK134609.1 | - | 4 | 964 | |||
AK140095.1 | - | 4 | 958 | |||
BC138255.1 | - | 4 | 390 | 8861274 | ||
BC138256.1 | - | 4 | 390 | 8861275 | ||
D43796.1 | - | 4 | 543 | |||
KC825358.1 | - | 3 | 352 | |||
U11763.1 | - | 4 | 548 | |||
U24699.1 | - | 3 | 361 | |||
U75372.1 | - | 4 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
