siRNA Details
Gene Symbol: Slc1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009201.2 | NP_033227.2 | 3 | 1186 | ||
GenBank mRNA | AF246130.1 | - | 3 | 631 | ||
AK148346.1 | - | 3 | 1186 | |||
AK157838.1 | - | 3 | 1165 | |||
AK182398.1 | - | 3 | 106 | |||
BC029873.1 | - | 3 | 689 | 4209657 | ||
BC037462.1 | - | 3 | 777 | 4192790 | ||
D85044.1 | - | 3 | 1119 | |||
L42115.1 | - | 3 | 1162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
