siRNA Details
Gene Symbol: Slc22a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011395.2 | NP_035525.1 | 4 | 1070 | ||
GenBank mRNA | AF078750.1 | - | 4 | 1070 | ||
AF082566.1 | - | 4 | 953 | |||
AK143988.1 | - | 4 | 740 | |||
BC137661.1 | - | 4 | 945 | 8860680 | ||
BC137662.1 | - | 4 | 945 | 8860681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
