siRNA Details
Gene Symbol: Slc22a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011396.3 | NP_035526.1 | 10 | 1785 | ||
GenBank mRNA | AB015800.1 | - | 10 | 1680 | ||
AF110417.1 | - | 10 | 1733 | |||
AF111425.1 | - | 10 | 1720 | |||
AK140640.1 | - | 10 | 1781 | |||
AK167451.1 | - | 10 | 1761 | |||
BC031118.1 | - | 10 | 1742 | 4981427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
