siRNA Details
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031197.2 | NP_112474.2 | 5 | 712 | ||
GenBank mRNA | AK005068.1 | - | 5 | 713 | ||
AK165430.1 | - | 5 | 647 | |||
BC034675.1 | - | 5 | 631 | 4236331 | ||
X15684.1 | - | 5 | 661 | |||
X16986.1 | - | 5 | 651 |

More Information
Other Species Targeted

Gene Ontology Categories:
