siRNA Details
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031197.2 | NP_112474.2 | 10 | 1299 | ||
GenBank mRNA | AK005068.1 | - | 10 | 1300 | ||
AK149532.1 | - | 3 | 412 | |||
AK165430.1 | - | 10 | 1234 | |||
BC034675.1 | - | 10 | 1218 | 4236331 | ||
S77926.1 | - | 2 | 38 | |||
X15684.1 | - | 10 | 1248 | |||
X16986.1 | - | 10 | 1238 |
