siRNA Details
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 4 | 882 | ||
GenBank mRNA | AK052623.1 | - | 4 | 853 | ||
AK132275.1 | - | 4 | 854 | |||
BC013441.1 | - | 4 | 824 | 4222972 | ||
D88533.1 | - | 4 | 842 |
