siRNA Details
Gene Symbol: Slc8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112798.2 | NP_001106269.1 | 2 | 1645 | ||
NM_001286684.1 | NP_001273613.1 | 2 | 1645 | |||
NM_011406.3 | NP_035536.2 | 2 | 1645 | |||
GenBank mRNA | AF004666.1 | - | 2 | 1640 | ||
AK048160.1 | - | 1212 | ||||
BC079673.1 | - | 1 | 1536 | 30532156 | ||
DQ388998.1 | - | 1 | 1535 | |||
U70033.1 | - | 2 | 1635 |
