siRNA Details
Gene Symbol: Slpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011414.3 | NP_035544.1 | 3 | 506 | ||
GenBank mRNA | AK010170.1 | - | 3 | 293 | ||
BC028509.1 | - | 3 | 506 | 1513866 | ||
U73004.1 | - | 717 | ||||
U88093.1 | - | 3 | 283 | |||
U94341.1 | - | 3 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
