siRNA Details
Gene Symbol: Snai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011415.2 | NP_035545.1 | 2 | 234 | ||
GenBank mRNA | AK133968.1 | - | 2 | 242 | ||
BC062164.1 | - | 2 | 234 | 30242610 | ||
U79550.1 | - | 2 | 162 | |||
U97059.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
