siRNA Details
Gene Symbol: Snai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011415.2 | NP_035545.1 | 2 | 504 | ||
GenBank mRNA | AK133968.1 | - | 2 | 512 | ||
BC062164.1 | - | 2 | 504 | 30242610 | ||
U79550.1 | - | 2 | 432 | |||
U97059.1 | - | 2 | 389 |

More Information
Other Species Targeted

Gene Ontology Categories:
