siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 20 | 2012 | ||
GenBank mRNA | AK077733.1 | - | 4 | 349 | ||
AK143485.1 | - | 20 | 2000 | |||
AK144122.1 | - | 20 | 1986 | |||
AK144631.1 | - | 20 | 1980 | |||
AK145352.1 | - | 20 | 1979 | |||
AK181148.1 | - | 3 | 189 | |||
AK199699.1 | - | 3 | 209 | |||
BC022173.1 | - | 20 | 1976 | |||
BC052423.1 | - | 20 | 1974 | 6400646 | ||
BC053064.1 | - | 20 | 1968 | |||
BC056375.1 | - | 20 | 1975 | |||
U85614.1 | - | 20 | 1970 |
