siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 16 | 1588 | ||
GenBank mRNA | AK143485.1 | - | 16 | 1576 | ||
AK144122.1 | - | 16 | 1562 | |||
AK144631.1 | - | 16 | 1556 | |||
AK145352.1 | - | 16 | 1555 | |||
AK204398.1 | - | 3 | 176 | |||
BC022173.1 | - | 16 | 1552 | |||
BC052423.1 | - | 16 | 1550 | 6400646 | ||
BC053064.1 | - | 16 | 1544 | |||
BC056375.1 | - | 16 | 1551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
