siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 6 | 707 | ||
GenBank mRNA | AK042194.1 | - | 7 | 620 | ||
AK133922.1 | - | 6 | 682 | |||
AK143485.1 | - | 6 | 695 | |||
AK144122.1 | - | 6 | 681 | |||
AK144631.1 | - | 6 | 675 | |||
AK145352.1 | - | 6 | 674 | |||
BC022173.1 | - | 6 | 671 | |||
BC052423.1 | - | 6 | 669 | 6400646 | ||
BC053064.1 | - | 6 | 663 | |||
BC056375.1 | - | 6 | 670 | |||
U85614.1 | - | 6 | 668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
