siRNA Details
Gene Symbol: Kdm5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013668.4 | NP_038696.2 | 11 | 1894 | ||
GenBank mRNA | AF127245.1 | - | 11 | 1849 | ||
AK129096.1 | - | 8 | 1068 | |||
AK155279.1 | - | 11 | 1879 | |||
AK155427.1 | - | 11 | 1879 | |||
AK155651.1 | - | 11 | 1892 | |||
AK158340.1 | - | 11 | 1879 | |||
BC043096.1 | - | 10 | 1726 | 6405943 | ||
BC054550.1 | - | 10 | 1726 | 6405943 | ||
Z29651.1 | - | 11 | 1456 |

More Information
Other Species Targeted

Gene Ontology Categories:
