siRNA Details
Gene Symbol: Smr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011422.3 | NP_035552.1 | 3 | 361 | ||
GenBank mRNA | BC031806.1 | - | 3 | 327 | 4192747 | |
BC031921.1 | - | 3 | 305 | 4912072 | ||
BC034553.1 | - | 3 | 331 | 4166033 | ||
BC035428.1 | - | 3 | 333 | |||
BC055857.1 | - | 3 | 355 | 4921509 | ||
BC141243.1 | - | 3 | 323 | 9055855 | ||
BC145569.1 | - | 3 | 323 | 9054134 | ||
BF454619.1 | - | 3 | 329 | |||
X71629.1 | - | 3 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
