siRNA Details
Gene Symbol: Ssty1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009220.2 | NP_033246.2 | 3 | 682 | ||
GenBank mRNA | X05260.1 | - | 3 | 682 |
Gene Symbol: Gm20831 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103152.1 | NP_001096622.1 | 733 | |||
GenBank mRNA | AK132684.1 | - | 734 | |||
AK161425.1 | - | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
