siRNA Details
Gene Symbol: Snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011432.2 | NP_035562.1 | 3 | 224 | ||
GenBank mRNA | AK137213.1 | - | 3 | 207 | ||
AK168973.1 | - | 3 | 197 | |||
BC008243.1 | - | 3 | 189 | 2655238 | ||
BC067386.1 | - | 1940 | ||||
BC092266.1 | - | 3 | 189 | 4009300 | ||
CJ085691.1 | - | 3 | 234 | |||
U70315.1 | - | 3 | 193 | |||
X96767.1 | - | 3 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
