siRNA Details
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 42 | 5853 | ||
GenBank mRNA | AB015790.1 | - | 42 | 5677 | ||
AF031816.1 | - | 5131 | ||||
AK088154.1 | - | 441 | ||||
AK147303.1 | - | 42 | 5854 | |||
AK169831.1 | - | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
