siRNA Details
Gene Symbol: Sorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011436.3 | NP_035566.2 | 31 | 4434 | ||
GenBank mRNA | AF031816.1 | - | 3712 | |||
AK137635.1 | - | 19 | 2549 | |||
AK147303.1 | - | 31 | 4435 | |||
Y12004.1 | - | 8 | 904 |

More Information
Other Species Targeted

Gene Ontology Categories:
