siRNA Details
Gene Symbol: Sox11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009234.6 | NP_033260.4 | 1 | 1261 | ||
GenBank mRNA | AB108675.1 | - | 1 | 951 | ||
AF009414.1 | - | 1 | 1239 | |||
AK146514.1 | - | 1 | 1221 | |||
AK165005.1 | - | 1 | 1260 | |||
AK165693.1 | - | 1 | 1256 | |||
BC053503.1 | - | 1 | 1117 | |||
BC062095.1 | - | 1 | 1064 | 5716171 | ||
BC078643.1 | - | 1 | 1246 | 30144974 |

More Information
Other Species Targeted

Gene Ontology Categories:
