siRNA Details
Gene Symbol: Sox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011448.4 | NP_035578.3 | 2 | 962 | ||
GenBank mRNA | AF421878.1 | - | 2 | 587 | ||
AK030187.1 | - | 2 | 947 | |||
AK031516.1 | - | 2 | 946 | |||
AK049986.1 | - | 2 | 948 | |||
AK220197.1 | - | 2 | 932 | |||
BC023796.1 | - | 2 | 949 | 5351850 | ||
BC023808.1 | - | 2 | 950 | 5354229 | ||
BC023953.1 | - | 2 | 959 | 5320371 | ||
BC024958.1 | - | 2 | 939 | 4949782 | ||
BC034264.1 | - | 2 | 890 | 4165469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
