siRNA Details
Gene Symbol: Spa17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011449.2 | NP_035579.1 | 3, 4 | 418 | ||
GenBank mRNA | AK005571.1 | - | 3, 4 | 287 | ||
BC059727.1 | - | 3, 4 | 288 | 6775177 | ||
BU936068.1 | - | 3, 4 | 423 | |||
CK137444.1 | - | 3, 4 | 279 | |||
Z46299.1 | - | 3, 4 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
