siRNA Details
Gene Symbol: Serpina1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009247.2 | NP_033273.1 | 3, 4 | 945 | ||
GenBank mRNA | AK004999.1 | - | 3, 4 | 947 | ||
AK135650.1 | - | 3, 4 | 950 | |||
AK146576.1 | - | 1147 | ||||
BC061176.1 | - | 3, 4 | 956 | 30381238 | ||
M75717.1 | - | 3, 4 | 940 |
Gene Symbol: Serpina1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252569.1 | NP_001239498.1 | 4, 5 | 1217 | ||
NM_009243.4 | NP_033269.1 | 3, 4 | 1002 | |||
GenBank mRNA | AK146619.1 | - | 935 | |||
BC011040.1 | - | 928 | 4159448 | |||
BC037007.1 | - | 1154 | 5123904 | |||
BC047128.1 | - | 928 | ||||
BC049970.1 | - | 928 | 4160332 | |||
BC057982.1 | - | 935 | 5134754 | |||
BC057984.1 | - | 950 | 5126441 | |||
BC057989.1 | - | 913 | 5100593 | |||
M75721.1 | - | 933 |
Gene Symbol: Serpina1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC009818.1 | - | 939 | 4211464 | ||
BC010984.1 | - | 939 | 4163602 | |||
BC010988.1 | - | 937 | ||||
BC011041.1 | - | 940 | 4193647 | |||
BC015266.1 | - | 891 | ||||
BC021325.1 | - | 980 | 5052898 | |||
BC021780.1 | - | 959 | 4160260 | |||
BC024108.1 | - | 950 | 5099206 | |||
BC031707.1 | - | 926 | 5123976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
