siRNA Details
Gene Symbol: Serpina3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011217.1 | - | 5 | 1249 | 4195356 | |
BC016407.1 | - | 5 | 1260 | 4210846 | ||
BC019802.1 | - | 5 | 1249 | 5050045 | ||
D00725.1 | - | 1250 | ||||
X55147.1 | - | 5 | 1255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
