siRNA Details
Gene Symbol: INCENP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040694.1 | NP_001035784.1 | 10 | 1760 | ||
NM_020238.2 | NP_064623.2 | 10 | 1760 | |||
XM_006718533.2 | XP_006718596.1 | 10 | 1722 | |||
XM_011544995.2 | XP_011543297.1 | 10 | 1722 | |||
XM_011544996.2 | XP_011543298.1 | 10 | 1722 | |||
XM_011544997.2 | XP_011543299.1 | 10 | 1682 | |||
XM_011544998.2 | XP_011543300.1 | 10 | 1722 | |||
GenBank mRNA | AF282265.1 | - | 9 | 1558 | ||
AK056195.1 | - | 10 | 1735 | |||
AK307633.1 | - | 10 | 1777 | |||
AY714053.1 | - | 9 | 1558 | |||
BC032678.1 | - | 10 | 1684 | |||
BC098576.1 | - | 10 | 1760 | 30924748 |
