siRNA Details
Gene Symbol: Serpina3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011458.2 | NP_035588.2 | 2 | 172 | ||
GenBank mRNA | AK004842.1 | - | 2 | 170 | ||
AK149436.1 | - | 2 | 173 | |||
BC011217.1 | - | 2 | 149 | 4195356 | ||
BC016407.1 | - | 2 | 160 | 4210846 | ||
BC019802.1 | - | 2 | 149 | 5050045 | ||
D00725.1 | - | 150 | ||||
X55147.1 | - | 2 | 155 | |||
X56786.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
