siRNA Details
Gene Symbol: Serpina3n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009252.2 | NP_033278.2 | 5 | 1289 | ||
GenBank mRNA | BC013651.1 | - | 5 | 1268 | 4160649 | |
M64086.1 | - | 5 | 1244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
