siRNA Details
Gene Symbol: Serpinb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009256.3 | NP_033282.1 | 5 | 663 | ||
GenBank mRNA | AK035624.1 | - | 5 | 602 | ||
AK087785.1 | - | 5 | 572 | |||
AK161581.1 | - | 5 | 664 | |||
BC029900.1 | - | 5 | 591 | 4925100 | ||
U96700.1 | - | 5 | 542 |
